missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Avidin, Texas Red™ conjugate

Product Code. 11544137
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 11544137

Brand: Invitrogen™ A820

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Avidin, Texas Red™ conjugate

Our avidin, Texas Red™ conjugate is a bright, photostable probe with excitation and emission maxima ∼595/615. Avidin is a glycosylated biotin-binding protein that has a basic isoelectric point and is widely used to detect biotinylated probes.
TRUSTED_SUSTAINABILITY

Specifications

Conjugate Texas Red
Quantity 5 mg
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Shipping Condition Room Temperature
Product Line Texas Red

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.