Learn More
SI Analytics™ AVS 370 Module 1 Viscometer
SI Analytics™ AVS 370 Module 1 Viscometer offers the extraordinary combination of both pressure and suction in operation.
Brand:  SI Analytics™ 1056516
Product Code. 15346511
  • 35660.00 € / Each
Estimated Shipment: 21-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Polymer 1
  • Scope of delivery: AVS 370 Basic unit, 1 pc. AVS/SK, 1 pc. VZ 8523, 1 pc. CT 53 HT, 1 pc. CK 300, 1 pc. VZ 8601, 1 pc. VZ 8604, 1 pc. VZ 8606, 1 pc. 8610, 1 pc. VZ 8630, 1 pc. VZ 8571, 1 pc. 8551, 1 pc. 8552
  • Gives you more flexibility and better adjustment to the liquids to be examined
  • It enables the operation of a Visco Pump II module for optical detection of transparent samples or the TC version for opaque and black fluids
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Viscometer
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
SI Analytics™ AVS 370 Module 1 Viscometer > Product Type: Viscometer