missing translation for 'onlineSavingsMsg'
Learn More

AW Standard Super-Cel™ NF Diatomaceous Earth, Honeywell Fluka™

Product Code. 15692150
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Suitable for most filtrations, derived from diatomaceous earth. Super Cel is a registered trademark of Imerys Minerals California, Inc.

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91053-39-3
Molecular Formula O2Si
Molecular Weight (g/mol) 60.083
MDL Number MFCD00132803
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name dioxosilane
SMILES O=[Si]=O

Specifications

Quantity 1 kg
Packaging Poly Bottle
UN Number NONH for all modes of transport
Chemical Name or Material Aw Standard Super-Cel™ Nf
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.