Learn More
Zeiss™ Axiolab 5 Upright Microscope for Transmitted Light Brightfield with an Ergonoimic Photo Binocular Tube
Compact, ergonomic upright microscope for use in the clinical environment and routine lab work. Configured for transmitted brightfield microscopy with a ergonomic photo binocular tube. Combine this microscope with the Axiocam 208 for SMART microscopy and easy digital documentation.
Brand:  Zeiss™ 490980-0006-000
Product Code. 17342820
  • 14105.00 € / 1 set
Estimated Shipment: 28-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Axiolab 5 is made for the routine microscopy work that goes on every day in your lab. Its compact and ergonomic design saves space and makes for easy handling.
  • Combine it with Axiocam 208 color for SMART microscopy. Just focus your sample and press a single button for crisp images in true color. The digital image will look like you see it through the eyepieces, with all the details and subtle color differences clearly visible. Plus, Axiolab 5 automatically adds the correct scaling information to your images. 
  • Microscope body is powder coated durable metal, heat treated and resistant to standard reagents used for staining, organic solvents used for cleaning of lenses and disinfectants.
  • All optical parts including objectives, eye pieces, lenses, prisms, have anti-reflective and anti-fungal coating.
  • Possible contrast methods are brightfield, darkfield, phase contrast, and simple transmitted light polarization. Upgrades from brightfield to other transmitted light contrasting techniques are available. With Axiolab 5 FL fluorescence is available in addition.
  • The microscope comes with a manual revolving nosepiece with 5 encoded positions, suitable for reading objective position for light intensity manager functionality and for scaling information
Specifications
Specifications
Transmitted Light Brightfield with an Ergonoimic Photobinocular Tube