missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 20 μL Ultra Micro Tips

Maxymum Recovery™ allows recovery of costly samples with improved precision and accuracy

28.50 € - 408.76 €

Specifications

Color Clear
Clean Claims DNase, RNase Free
For Use With (Application) Allows recovery of costly samples with improved precision and accuracy
Tip Style Micro Point
For Use With (Equipment) Gilson™ Pipetman™ P10, Gilson™ Pipetman™ P2, Eppendorf Pipetter
View More Specs

Products 4
Product Code Brand Sterility Format Price Quantity & Availability  
Product Code Brand Sterility Format Price Quantity & Availability  
12146880
View Documents
Axygen™
T-400
Non-sterile Bulk
28.50 €
Pack of 1000
In Stock
to see stock.
Edge
Add to basket
 
12541368
View Documents
Axygen™
T-400
Non-sterile Bulk This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12726619
View Documents
Axygen™
T-400-R-S
Sterile Rack This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12716619
View Documents
Axygen™
T-400-R
Non-sterile Rack This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors, compatible with most popular brands of pipettors. These polypropylene tips are nonpyrogenic and RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.

  • RNase-/DNase-free
  • Nonpyrogenic
  • Maxymum Recovery technology for ultra-low retention

Order Info

  • Bulk: 1000 tips/pack, 20 packs/case
  • Racked: 96 tips/rack, 10 racks/pack, 5 packs/case

Specifications

Specifications

Clear
Allows recovery of costly samples with improved precision and accuracy
Gilson™ Pipetman™ P10, Gilson™ Pipetman™ P2, Eppendorf Pipetter
DNase, RNase Free
Micro Point
Polypropylene
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Axygen™ 20 μL Ultra Micro Tips

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.