Learn More
Axygen™ 1000 μL Universal Pipetter Tips: Bevelled
GREENER_CHOICE
Compatible with most popular brands of pipettes
Brand:  Axygen™ T-1000-B
21.20 EUR valid until 2025-03-30
Use promo code "24134" to get your promotional price.
 View more versions of this product
Product Code. 12176870
  • 33.10 € / Pack of 1000
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors, compatible with most popular brands of pipettors. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.
  • These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
  • Nonpyrogenic
  • Beveled
TRUSTED_SUSTAINABILITY
Specifications
Specifications
100 to 1000 μL
Blue
DNase, RNase, and Pyrogen free
Beveled, Universal
Universal Fit
Non-sterile
Autoclavable
Bulk
1,000 Tips/Pk., 5 Packs/Cs.
Polypropylene
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Axygen™ 1000 μL Universal Pipetter Tips: Bevelled > Bulk; Standard; 1-1,000 μL