missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 200 μL Universal Pipetter Tips: 200 μL, Wide Bore

Designed to fit a wide variety of single and multi-channel pipettes

30.65 € - 427.25 €

Specifications

Volume (Metric) 1 to 200 μL
Color Clear
Autoclavable Autoclavable
Clean Claims DNase, RNase, and Pyrogen free
For Use With (Equipment) Universal Fit Pipettor
View More Specs

Products 3
Product Code Brand Sterility Price Quantity & Availability  
Product Code Brand Sterility Price Quantity & Availability  
11396054
View Documents
Axygen™
T-205-WB-C-R-S
Sterile
408.00 €
Pack of 4800
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
 
12581358
View Documents
Axygen™
T-205-WB-C
Non-sterile This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11374245
View Documents
Axygen™
T-205-WB-C
Non-sterile
30.65 €
Pack of 1000
In Stock
to see stock.
Add to basket
 
Description

Description

Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.

(This is the suggested replacement for T-205-WB-C-L)

  • These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
  • Nonpyrogenic
  • Wide-Bore

Order Info

Available in bulk, racked or stack rack formats.

Specifications

Specifications

1 to 200 μL
Autoclavable
Universal Fit Pipettor
Clear
DNase, RNase, and Pyrogen free
Polypropylene
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.