missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 384-well Skirted PCR Microplates
GREENER_CHOICE

Single and double notch 384-well PCR plates

168.48 € - 983.87 €

Specifications

Certifications/Compliance RNase, DNase-free
Format Standard
Array 16 x 24
Material Polypropylene
No. of Wells 384
View More Specs

Products 11
Produktcode Marke Color Volume (Metric) Well Includes Preis Menge & Verfügbarkeit  
Produktcode Marke Color Volume (Metric) Well Includes Preis Menge & Verfügbarkeit  
12719418
missing translation for 'viewDocuments'
Axygen™
PCR-384-B
Blue Double notch This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12749418
missing translation for 'viewDocuments'
Axygen™
PCR-384-C-S
Clear 50 μL Single notch This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12759418
missing translation for 'viewDocuments'
Axygen™
PCR-384-G
Green Double notch This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12709438
missing translation for 'viewDocuments'
Axygen™
PCR-384-O
Orange Double notch This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10068469
missing translation for 'viewDocuments'
Axygen™
PCR-384-LC480-W-NF
White 55 μL Single notch
339.81 €
Pack of 50
Estimated Shipment: 02-06-2025
to see stock.
Zum Warenkorb hinzufügen
 
12739438
GREENER_CHOICE
missing translation for 'viewDocuments'
Axygen™
PCR-384-RGD-C
Clear 50 μL Double notch
983.87 €
Pack of 100
Estimated Shipment: 31-03-2025
to see stock.
Zum Warenkorb hinzufügen
 
12567817
GREENER_CHOICE
missing translation for 'viewDocuments'
Axygen™
PCR-384-RGD-C
Clear 50 μL Double notch This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12719438
missing translation for 'viewDocuments'
Axygen™
PCR-384-R
Red 40 μL Double notch This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12759438
missing translation for 'viewDocuments'
Axygen™
PCR-384-V
Violet 40 μL Double notch This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12769438
missing translation for 'viewDocuments'
Axygen™
PCR-384-Y
Yellow Double notch This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15328301
GREENER_CHOICE
missing translation for 'viewDocuments'
Axygen™
PCR-384M2-C-PBC
Clear 50 μL Single notch
429.97 €
Pack of 50
Estimated Shipment: 02-06-2025
to see stock.
Zum Warenkorb hinzufügen
 
Beschreibung

Beschreibung

Sealing Options: Axymats, heat sealing, adhesive films and foils.

  • Available in 50μL and 55μL
  • Single and double notch corner

Skirted (50μL)

  • Application/Compatibility: standard thermal cycling
  • Robotic-friendly full skirt design
  • Packaging Description: 10 plates/Pk., 5 packs/Cs.
Skirted (55μL)
  • Application/Compatibility: standard thermal cycling, fully ABI compatible for qPCR and sequencing
  • Robotic-friendly full skirt design
  • Packaging Description: 10 plates/Pk., 5 packs/Cs.
Skirted, RigidPlate (50μL)
  • Robotic-friendly rigid plate with full skirt
  • Application/Compatibility: standard thermal cycling, ABI sequencing, automation applications
  • Packaging Description: 10 plates/Pk., 2 packs/sleeve, 5 sleeves/Cs.
Skirted, elevated wells
  • Application/Compatibility: standard thermal cycling, ABI sequencing, qPCR
  • Raised wells allow 55μL volume
  • Packaging Description: 10 plates/Pk., 5 packs/Cs.

Colors available by special order.

Spezifikation

Spezifikation

RNase, DNase-free
16 x 24
384
Standard
Polypropylene
Round
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt