missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ 48-Well PCR Microplates
Specially designed to fit most thermal cyclers
Brand: Axygen™ PCR-48-C
This item is not returnable.
View return policy
Description
- Volume to brim of well: 350 μL, working volume: 300 μL, volume with AxyMat: 240 μL
Specifications
RNase, DNase, DNA-free | |
Without Lid | |
Clear | |
48 | |
Round | |
50/Cs. |
Standard | |
6 x 8 | |
Polypropylene | |
Non-sterile | |
PCR | |
10/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction