missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 96-well PCR Microplates

Product Code. 12739458 Shop All Axygen Scientific Products
Click to view available options
Color:
Assorted
Black
Blue
Clear
Orange
Red
White
Yellow
Skirt Style:
Elevated
Full
Half
Unskirted
Format:
Elevated Wells
Flat Top
Single Notch Corner
Standard
Unit Size:
Pack of 10
Pack of 100
Pack of 25
Pack of 50
This item is not returnable. View return policy

Product Code. 12739458

Brand: Axygen™ PCR96FSO

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Constructed with ultra-thin and consistent wall thickness to allow precise thermal transfer for optimum results. Axygen™ 96-well PCR Microplates are the ideal option for use in PCR thermal transfer and Real-Time PCR applications.

Axygen™ 96 well elevated skirt PCR microplate is designed to fit ABI Real Time PCR thermal cyclers and sequencers. Ultra-thin and consistent wall thickness allows precise thermal transfer for optimum results. Can be sealed with 8/12 strip caps, Axymat or sealing film.


  • RNase-/DNase-free
  • Nonpyrogenic
  • Nonsterile
  • Manufactured with 99.9% pure virgin polypropylene
  • 96-well Semi-Skirted PCR Plate
  • Single Notch Coner
  • Application/Compatibility: Standard thermal cycling, ABI Real time PCR 7300, 7500, 7900, qPCR and ABI Sequencing
  • TRUSTED_SUSTAINABILITY

    Specifications

    Barcode No Barcode
    Certifications/Compliance RNase-, DNase-free
    Format Standard
    Array 8 x 12
    Color Orange
    For Use With (Equipment) Standard 96 well blocks
    Skirt Style Full
    Material Polypropylene
    Volume (Metric) Well 200 μL
    No. of Wells 96
    Sterility Non-sterile
    Well Shape Round
    For Use With (Application) PCR thermal transfer, RT-PCR applications
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.