Learn More
Axygen™ AxyMats™ 96 Round Well Expansion Mat for PCR Microplates
Suited to help reduce evaporation and ensure proper seal when used in conjunction with Axygen PCR Microplates and PlateMax™ sealing films
Brand: Axygen CM-96-EXP
This item is not returnable.
View return policy
Description
- Made of research-grade silicone
- Autoclavable and re-usable
- For use with 96/384 well and PCR microplates
- Non-sterile
Specifications
Natural | |
Non-sterile | |
96 | |
10 | |
Round |
Silicone | |
Autoclavable | |
5 | |
Expansion Mat | |
96/384-well PCR Microplate |