Learn More
Axygen™ AxyMats™ Sealing Mat for 750uL 96 Round Well Plate and ELISA Plate
Minimize evaporation in wide range of temperatures in PCR and storage applications
Brand:  Axygen™ AM-750UL-RD
Product Code. 12567897
  • 331.99 € / Pack of 50
Estimated Shipment: 23-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Axygen AxyMat sealing mats can be used in a wide range of temperatures for PCR and storage applications. AxyMats minimize evaporation and are made of research-grade silicone which can be autoclaved. They can be reused following a bleach wash and ethanol rinse protocol.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Natural
Non-sterile
96
10/Pk.
Round
Silicone
Autoclavable
50/Cs.
Sealing Mat
96-well Round Well Plate and ELISA Plate, 750 μL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Axygen™ AxyMats™ Sealing Mat for 750uL 96 Round Well Plate and ELISA Plate > No; 96-well; Round