Learn More
Axygen™ Conical Centrifuge Tubes
GREENER_CHOICE
Graduated tubes with conical bottom
Brand:  Axygen™ SCT-15ML-500
145.57 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 11324175
  • 159.00 € / Pack of 500
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Rated to 12,000 RCF
  • Printed graduations and writing area for sample identification
  • Screw caps can be closed with one hand.
  • Sterile options are also available.
  • RNase-/DNase-free
  • Nonpyrogenic
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Screw Cap
Not Autoclavable
Clear
For sample identification
25 Tubes and caps/Rack; 20 Racks/Cs.
Conical
12,000 x g
15 mL
Polypropylene
Screw cap
Non-sterile