missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Horizontal Gel Box, 10 cm

The Axygen HGB-10 is a compact 10cm wide gel box capable of running up to 100 samples depending on configuration. System is ideal for quick sample-checks, particularly following restriction digestion during cloning. Comes with two different size UV transparent gel trays providing flexibility and value.

Brand:  Axygen™ HGB-10

489.53 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


Product Code. 15509923

  • 535.00 € / Each
Estimated Shipment: 15-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Axygen horizontal gel boxes are high-quality, full-featured units designed for exceptional durability and efficiency. The gel boxes feature leak-free external gel casting, which allows you to cast one gel outside of the box while another gel is running inside. The units are available in four different sizes, and each unit comes with two sizes of gel trays. In addition, they have easy to replace electrodes that are recessed to reduce potential damage. These gel boxes are an excellent value that will provide years of reliable service.



Features
  • Flexibility – Each unit comes with 2 sizes of gel trays to optimize applications
  • External gel casting to gel box allows for running gels while casting the next one
  • Ease of use – the Axygen contrast enhancer makes loading gels easier
  • Rugged injection molding is supported by a 3-year warranty
  • Larger units come with recirculation ports allowing for buffer exchange

Four sizes of horizontal tanks. Each comes with 2 sizes of gel trays which allow for external gel casting. Three larger sizes come with recirculation ports for use as needed.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CE certified
1/Cs.
300 mL
10 cm
Horizontal Gel Box
1/Pk.
12.5 x 22 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers