Learn More
Axygen™ Horizontal Gel Box, 15 cm
The Axygen HGB-15 is large sample capacity system ideal for screening PCR samples or routine DNA fragment analysis and comes standard with two different size UV transparent gel trays providing flexibility and value.
Brand: Axygen™ HGB-15
737.82 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
Description
Axygen horizontal gel boxes are high-quality, full-featured units designed for exceptional durability and efficiency. The gel boxes feature leak-free external gel casting, which allows you to cast one gel outside of the box while another gel is running inside. The units are available in four different sizes, and each unit comes with two sizes of gel trays. In addition, they have easy to replace electrodes that are recessed to reduce potential damage. These gel boxes are an excellent value that will provide years of reliable service.
Features
- Flexibility – Each unit comes with 2 sizes of gel trays to optimize applications
- External gel casting to gel box allows for running gels while casting the next one
- Ease of use – the Axygen contrast enhancer makes loading gels easier
- Rugged injection molding is supported by a 3-year warranty
- Larger units come with recirculation ports allowing for buffer exchange
Four sizes of horizontal tanks. Each comes with 2 sizes of gel trays which allow for external gel casting. Three larger sizes come with recirculation ports for use as needed.
Specifications
CE certified | |
1/Cs. | |
500 mL | |
15 cm |
Horizontal Gel Box | |
1/Pk. | |
17.5 x 26.5 cm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.