missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Microplate Sealing Film and Tapes

Offer great economy for disposable ease.

Brand:  Axygen™ UC-500

214.82 EUR valid until 2025-04-13
Use promo code "24122" to get your promotional price.


 View more versions of this product

Product Code. 11304325

  • 361.00 € / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

For applications ranging from PCR and Real-Time PCR to ELISA and cell culture, Axygen films are an easy and cost-effective way to seal plates and enhance automation. Used to seal multi-well microplates. Sealing film temperature ranges from -40°C to +104°C.

Breathable

  • Allow effective gas exchange for cellular and bacterial cultivation — while preventing contamination
  • Seal polypropylene and polystyrene culture plates, 96- and 384-well plates including other assay plates
Aluminum
  • Provides exceptional seal on multi-well plates with varied temperatures and reagents
  • With compression mats, films and tapes seal during thermocycling
  • Ideal for light-sensitive samples and its uniformly applied adhesive provides optimum well sealing
  • Pierceable, PCR-AS-200
CyclerSeal
  • Exceptional seal across entire plate safeguards samples during transport or storage
  • When used with Axygen's compression mat, this polyester film eliminates well-to-well contamination and cross-over in PCR applications
AxySeal
  • Polyester-based with uniformly and consistently applied acrylic adhesive to eliminate the edge effect in sensitive ELISA assays
  • Suitable for sealing tissue culture plates, short-term storage and incubation, transport and the containment of biohazardous solutions
  • Available in both nonsterile and sterile packaging

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Polyolefin
500/Cs.
Not Available
UltraClear Sealing Film
-80°C to +104°C
Non-sterile
100/Pk.
Designed for applications ranging from PCR and real-time PCR to ELISA and cell culture, film clarity allows for optical analysis during real-time PCR
70 μm
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Axygen™ Microplate Sealing Film and Tapes > Polyolefin; 97 μm; -70 °C - 100 °C; Real-Time PCR

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.