missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Presterilized Conical Centrifuge Tubes
GREENER_CHOICE

Graduated tubes with conical bottom

163.63 € - 219.34 €

Specifications

Cap Style Screw Cap
Autoclavable Not Autoclavable
Color Clear
Material Polypropylene
For Use With (Application) For sample identification
View More Specs

Products 4
Product Code Brand Capacity (Metric) Packaging Price Quantity & Availability  
Product Code Brand Capacity (Metric) Packaging Price Quantity & Availability  
12569067
GREENER_CHOICE
View Documents
Axygen™
SCT-15ML-R-S
15 mL 50 Tubes and Caps/Unit, 20 Units/Cs.
219.34 €
Pack of 500
Estimated Shipment: 14-03-2025
to see stock.
Add to basket
 
12519107
GREENER_CHOICE
View Documents
Axygen™
SCT-50ML-25-S
50 mL 25 Tubes and Caps/Unit, 20 Units/Cs.
163.63 €
Pack of 500
Estimated Shipment: 14-03-2025
to see stock.
Add to basket
 
12549067
GREENER_CHOICE
View Documents
Axygen™
SCT-15ML-25-S
15 mL Bag
194.39 €
Pack of 500
Estimated Shipment: 14-03-2025
to see stock.
Add to basket
 
12539107
GREENER_CHOICE
View Documents
Axygen™
SCT-50ML-R-S
50 mL 25 Tubes and Caps/Rack, 20 Racks/Cs.
183.29 €
Pack of 500
Estimated Shipment: 18-07-2025
to see stock.
Add to basket
 
Description

Description

  • Rated to 12,000 RCF
  • Printed graduations and writing area for sample identification
  • Screw caps can be closed with one hand.
  • Sterile options are also available.
  • RNase-/DNase-free
  • Nonpyrogenic
Specifications

Specifications

Screw Cap
Clear
For sample identification
Sterile
Not Autoclavable
Polypropylene
Screw cap
12,000 x g
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.