Learn More
Axygen™ Screw Cap Tubes without Caps: Self-Standing
Self-standing tubes
Brand:  Axygen™ ST-050-SS-S
 View more versions of this product
Product Code. 12779598
  • 780.00 € / Pack of 4000
Estimated Shipment: 13-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Axygen microcentrifuge tubes feature a screw cap that provides a tight secure seal. Screw cap tubes are available in a variety of colors with a conical or self-standing bottom. Sterile options are also available.
  • Maximum G-force is 20,000 RCF.
  • RNase-/DNase-free
  • Nonpyrogenic
Tubes, with Cap, Sterile
All sterile screw cap microcentrifuge tubes are pre-assembled with non-looped caps with O-Rings.
Colored-cap versions are also available in sterile format.
Order Info
Caps sold separately.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RNase, DNase, pyrogen-free
0.5 mL
Polypropylene
Sterile
Not Included
20,000 x g
Clear
Bag
No
Conical