missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Screw Caps with O-rings

Select looped or non-looped format

Brand:  Axygen™ SCO-LP-BR

 View more versions of this product

Product Code. 12769378

  • 810.00 € / Pack of 4000
Estimated Shipment: 13-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Axygen microcentrifuge tubes feature a screw cap that provides a tight secure seal. Screw cap tubes are available in a variety of colors with a conical or self-standing bottom. Sterile options are also available.

  • Maximum G-force is 20,000 RCF.
  • RNase-/DNase-free
  • Nonpyrogenic


Tubes, with Cap, Sterile


All sterile screw cap microcentrifuge tubes are pre-assembled with non-looped caps with O-Rings.


Colored-cap versions are also available in sterile format.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

O-ring
Brown
500/Pk.
Non-sterile
Polypropylene
Looped
4000/Cs.
Both skirted and non-skirted variations of the 0.5 mL, 1.5 mL, and 2 mL microcentrifuge tubes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers