missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Storage Boxes

Axygen™ Storage Boxes can store samples for an extended period of time.

Brand:  Axygen™ TR8300-BLK

 View more versions of this product

Product Code. 16535954

  • 39.49 € / Pack of 2

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Supports freezing at -80°C and autoclaving at 121°C
  • Alphanumeric Sample identification
  • Engieneered to simplify sample identity
  • Polypropylene structure is ensured to last
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Sample size: 1.5/2.0 mL
Black Opaque
Sample Storage
Storage Box
-80°C to +121°C
14.2 cm
Autoclavable
Polypropylene
100 Tubes
Case of 2
14.2 cm
5.5 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions