missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Storage Microplates
For sample collection, in-vitro growth chambers and long term storage.
Brand: Axygen™ P-96-450V-C-S
154.88 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
Axygen™ deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage. Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to -80°C.
- 450 μL deep well plate
- ANSI/SLAS 1-2004: Microplates - Footprint Dimensions
- Ultra-flat surface ensures proper sealing with heat sealing film
Specifications
ANSI, SLAS 1-2004 | |
Without Lid | |
Autoclavable | |
96-well Storage V-bottom Microplate | |
550 μL | |
Sterile | |
Sample collection, in-vitro growth chambers and long term storage | |
50/Cs. | |
-112°F |
Standard | |
8 x 12 | |
Clear | |
Polypropylene | |
96 | |
Conical Bottom (V-well) | |
4000 x g | |
10/Pk. | |
-80°C |