missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Storage Microplates
BIOPHARMAGREENER_CHOICE

For sample collection, in-vitro growth chambers and long term storage.

Brand:  Axygen™ P-2ML-SQ-C-S

139.89 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.


Product Code. 11370105

  • 177.90 € / Pack of 25
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Axygen™ deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage. Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to -80°C.


  • 2.2 mL deep well plate
  • ANSI/SLAS 1-2004: Microplates - Footprint Dimensions
  • Ultra-flat surface ensures proper sealing with heat sealing film
  • TRUSTED_SUSTAINABILITY
    Specifications

    Specifications

    ANSI, SLAS 1-2004
    Without Lid
    Clear
    96
    Square
    25/Cs.
    -112°F
    Standard
    8 x 12
    Polypropylene
    Sterile
    Sample collection, in-vitro growth chambers and long term storage
    25/Pk.
    -80°C
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents Certificates

    Certificates

    A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

    5 results found
    Lot Number Certificate Type Date Catalog Number
    Lot Number24524013 Certificate TypeCertificate of Quality Date08/01/2025 Catalog NumberP-2ML-SQ-C-S
    Lot Number23224080 Certificate TypeCertificate of Quality Date01/01/2025 Catalog NumberP-2ML-SQ-C-S
    Lot Number24524021 Certificate TypeCertificate of Quality Date25/12/2024 Catalog NumberP-2ML-SQ-C-S
    Lot Number24524014 Certificate TypeCertificate of Quality Date25/12/2024 Catalog NumberP-2ML-SQ-C-S
    Lot Number23124016 Certificate TypeCertificate of Quality Date18/12/2024 Catalog NumberP-2ML-SQ-C-S
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title
    Axygen™ Storage Microplates > 96-well; 2.2 mL; Yes; Square

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.