missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Wellpro™ Robotic Tips
Clear tips
484.71 € - 494.34 €
Specifications
Volume (Metric) | 200 μL |
---|---|
Filtered | Non-filtered |
Color | Clear |
Rack Color | Gray |
Certifications/Compliance | Non-ANSI |
Description
- Racked
- 200μL capacity
- Presterilized tips available
- Grey racks
Order Info
- 30++l
- 200++l
Specifications
200 μL | |
Clear | |
Non-ANSI | |
Non-pyrogenic, used for stringent specifications under strict process controls | |
Automation, Round | |
Polypropylene |
Non-filtered | |
Gray | |
DNase, Rnase Free | |
Rack | |
ProGroup Wellpro 3000 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title