Learn More
Axygen™ Axypet™ 8-Channel Digital Pipettor, Calibration 3x10 STD
High levels of accuracy and precision with design innovations to make pipetting more comfortable during prolonged periods of time.
795.00 €
Specifications
Certifications/Compliance ISO17025
Number of Channels 8
Product Line Axypet™
Type Multi-channel Pipettor
Quantity 1/Pk., 1/Cs
View More Specs
Products 4
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
17223634
View Documents
Axygen™
AP-8-10-STD
0.5 to 10 μL
795.00 €
Each
In Stock
to see stock.
Add to basket
17283634
View Documents
Axygen™
AP-8-50-STD
5 to 50 μL
795.00 €
Each
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
17243634
View Documents
Axygen™
AP-8-200-STD
20 to 200 μL
795.00 €
Each
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
17263634
View Documents
Axygen™
AP-8-300-STD
50 to 300 μL
795.00 €
Each
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
Description
Description
  • ISO 17025 Pre-calibrated
  • Thumb-friendly soft spring pipetting system reduces fatigue
  • Unique side EZIject button option reduces thumb movement
  • Finger-hook provides for a more relaxed grip
  • Large ergonomic push button is easy on the thumb during contact
  • Locking feature prevents accidental volume changes during pipetting procedures
  • Push button and ejector button are ergonomically located
  • Curved ejector bar allows tips to be ejected in steps, reducing the mount of force required for ejection
  • Tip manifold rotates 360°
  • Each channel of the pipettor has an individual, precision piston assembly to ensure accuracy and reproducibility from one pipetting series to the next
  • Shafts withdraw when pressed against a row of tips
  • Each shaft moves independently, minimizing jamming of tips
  • Micrometer is continuously adjustable for selection of whole or fractional volumes
Specifications
Specifications
ISO17025
Axypet™
1/Pk., 1/Cs
8
Multi-channel Pipettor