missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Axypet™ Single-Channel Pipettor
![GREENER_CHOICE](/content/dam/fishersci/glyphs/green_glyph.png)
Axygen™ Axypet™ pipettors combine high levels of accuracy and precision with design innovations to make pipetting more comfortable during prolonged periods of time.
Brand: Axygen™ AP-250
This item is not returnable.
View return policy
Description
- Thumb-friendly soft spring pipetting system reduces fatigue
- 3-year warranty
- Unique side EZIject button option reduces thumb movement
- Finger-hook provides for a more relaxed grip
- Large ergonomic push button is easy on the thumb during contact
- Locking feature prevents accidental volume changes during pipetting procedures
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
±1.0/ ±0.6% | |
1 | |
Single-Channel Pipettor | |
±0.4/ ±0.3% |
Tip Type: 300 μL | |
Axypet | |
50 to 250 μL | |
3 years |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction