Learn More
Axygen™ Axypet™ Single Channel Pipettor, Calibration 3x10 STD
High levels of accuracy and precision with design innovations to make pipetting more comfortable during prolonged periods of time.
252.27 € - 278.42 €
Specifications
Certifications/Compliance | ISO17025 |
---|---|
Number of Channels | 1 |
Product Line | Axypet™ |
Type | Single Channel Pipettor |
Quantity | 1/Pk., 1/Cs |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
17203624
|
Axygen™
AP-2-STD |
0.1 to 2 μL |
252.27 €
Each |
In Stock
Log in to see stock. |
|||||
17243604
|
Axygen™
AP-10-STD |
0.5 to 10 μL |
252.27 €
Each |
Estimated Shipment: 03-06-2025
Log in to see stock. |
|||||
17203634
|
Axygen™
AP-5000-STD |
1 to 5 mL |
278.42 €
Each |
Estimated Shipment: 03-06-2025
Log in to see stock. |
|||||
17203614
|
Axygen™
AP-10ML-STD |
1 to 10 mL |
278.42 €
Each |
Estimated Shipment: 03-06-2025
Log in to see stock. |
|||||
17223624
|
Axygen™
AP-20-STD |
2 to 20 μL |
252.27 €
Each |
Estimated Shipment: 03-06-2025
Log in to see stock. |
|||||
17283624
|
Axygen™
AP-50-STD |
5 to 50 μL |
252.27 €
Each |
Estimated Shipment: 03-06-2025
Log in to see stock. |
|||||
17263604
|
Axygen™
AP-100-STD |
10 to 100 μL |
252.27 €
Each |
Estimated Shipment: 03-06-2025
Log in to see stock. |
|||||
17243624
|
Axygen™
AP-200-STD |
20 to 200 μL |
252.27 €
Each |
In Stock
Log in to see stock. |
|||||
17263624
|
Axygen™
AP-250-STD |
50 to 250 μL |
252.27 €
Each |
Estimated Shipment: 03-06-2025
Log in to see stock. |
|||||
17283604
|
Axygen™
AP-1000-STD |
100 to 1000 μL |
252.27 €
Each |
Estimated Shipment: 03-06-2025
Log in to see stock. |
|||||
Description
- ISO 17025 Pre-calibrated
- Thumb-friendly soft spring pipetting system reduces fatigue
- Unique side EZIject button option reduces thumb movement (Left handed EZIject button available by special order).
- Finger-hook provides for a more relaxed grip
- Large ergonomic push button is easy on the thumb during contact
- Locking feature prevents accidental volume changes during pipetting procedures
Specifications
ISO17025 | |
Axypet™ | |
1/Pk., 1/Cs |
1 | |
Single Channel Pipettor |