Learn More
Axygen™ Axypet™ Starter Kit
GREENER_CHOICE
CORNING AXYGEN™ Axypet™ Starter Kit includes a set of micropipettes, tips, and other quality accessories.
Brand:  Axygen™ AP-STR-KIT-1
502.78 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 15753957
  • 540.00 € / 1 set
Estimated Shipment: 16-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Micropipette starter kit
  • Includes micropipettes, tips, calibration tools, identification tools, and more
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Axypet
20 to 1000 μL
1 Kit
Starter Kit, Micro
AP-20, AP-200, AP-1000 micropipettes, 1 rack each of 200μL and 1000μL tips, a calibration tool, ejector spacers, identification labels, lubricant, color identification rings, right-hand ejector button