missing translation for 'onlineSavingsMsg'
Learn More

AZD 2461, Tocris Bioscience™

Product Code. 30000999
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent PARP inhibitor; orally bioavailable

AZD 2461 is a potent PARP inhibitor (IC50 values are 2, 5 and 200 nM for PARP2, PARP1 and PARP3, respectively). Exhibits anticancer effects in BRCA1 mutant, but not wild-type breast cancer cell lines in vitro. Inhibits growth of olaparib -resistant mammary tumors in a mouse model and is a poor substrate for the P-gp transporter. Orally bioavailable.

Chemical Identifiers

CAS 1174043-16-3
Molecular Formula C22H22FN3O3
Molecular Weight (g/mol) 395.434
InChI Key HYNBNUYQTQIHJK-UHFFFAOYSA-N
Synonym AZD 2461
PubChem CID 44199317
IUPAC Name 4-[[4-fluoro-3-(4-methoxypiperidine-1-carbonyl)phenyl]methyl]-2H-phthalazin-1-one
SMILES COC1CCN(CC1)C(=O)C2=C(C=CC(=C2)CC3=NNC(=O)C4=CC=CC=C43)F

Specifications

Linear Formula C22H22FN3O3
Quantity 50 mg
Solubility Information Soluble to 100mM in DMSO and to 100mM in ethanol
Formula Weight 395.43
Physical Form Solid
Chemical Name or Material 4-[[4-Fluoro-3-[(4-methoxy-1-piperidinyl)carbonyl]phenyl]methyl]-1(2H)-phthalazinone
Grade >98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.