Learn More
Tocris Bioscience™ AZD 5582 dihydrochloride
Dimeric Smac mimetic; potent IAP inhibitor
Brand:  Tocris Bioscience™ 5141/10
232.39 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18709017
  • 294.00 € / 10mg
Explore more special offers
Description
Description
AZD 5582 dihydrochloride is a dimeric Smac mimetic; potent inhibitor of X-linked (XIAP) and cellular (cIAP) inhibitor of apoptosis protein (IC50 values are 15, 15 and 21 nM for XIAP, cIAP1 and cIAP2 respectively). Binds to the BIR3 domain of XIAP to prevent interaction with caspase-9. Causes degradation of cIAP1 and cIAP2 and induces apoptosis in MDA-MB-231 breast cancer cells. Causes tumor regression in MDA-MB-231 xenograft-bearing mice.
Specifications
Specifications
>97%
AZD 5582 dihydrochloride
AZD 5582 dihydrochloride
10 mg
1088.21