missing translation for 'onlineSavingsMsg'
Learn More

Azidotri-n-butyltin(IV), 95%, Thermo Scientific Chemicals

Product Code. 15421948
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Azidotri-n-butyltin(IV) is widely utilized as a reagent in the synthesis of tetrazolylbenzene compounds. It is employed in pharmaceutical research and involves synthesis of tetrazole derivatives, which is used to create angiotensin II receptor antagonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Azidotri-n-butyltin(IV) is widely utilized as a reagent in the synthesis of tetrazolylbenzene compounds. It is employed in pharmaceutical research and involves synthesis of tetrazole derivatives, which is used to create angiotensin II receptor antagonists.

Solubility
Miscible with toluene, hexane, acetonitrile and tetrahydrofuran.

Notes
Use in a fume hood. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17846-68-3
Molecular Formula C12H27N3Sn
Molecular Weight (g/mol) 332.079
MDL Number MFCD00216557
InChI Key JKVRTUCVPZTEQZ-UHFFFAOYSA-N
Synonym tributyltin azide, azidotributyltin iv, azidotributyltin, azidotributyl tin, tri-n-butylazidotin, azido tributyl stannane, azido tributyltin, tri-butyltin azide, tributyl tin azide, tributylstannylazide
PubChem CID 4984872
IUPAC Name azido(tributyl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)N=[N+]=[N-]

Specifications

Density 1.212
Flash Point 113°C (235°F)
Linear Formula [CH3(CH2)3]3SnN3
Quantity 1 g
UN Number UN2788
Solubility Information Miscible with toluene,hexane,acetonitrile and tetrahydrofuran.
Formula Weight 332.06
Percent Purity 95%
Chemical Name or Material Azidotri-n-butyltin(IV)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'