missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azithromycin
Click to view available options
Quantity:
50 mg
Description
Azithromycin is a macrolide antibiotic. Inhibits 50S ribosomal subunit formation and elongation at transpeptidation step in gram-positive and gram-negative organisms. Orally active with improved pharmacokinetics over erythromycin in mouse models. Inhibits autophagy. Predicted to disrupt binding of SARS-CoV-2 spike protein to ACE2.
Specifications
Specifications
Type | Azithromycin |
CAS | 83905-01-5 |
Quantity | 50 mg |
Molecular Formula | C38H72N2O12 |
MDL Number | MFCD00873574,MFCD01862248,MFCD03191387 |
Synonym | 13-[(2,6-Dideoxy-3-C-methyl-3-O-methyl-α-L-ribo-hexopyranosyl)oxy]-2-ethyl-3,4,10-trihydroxy-3,5,6,8,10,12,14-heptamethyl-11-[[3,4,6-trideoxy-3-(dimethylamino)-β-D-xylo-hexopyranosyl]oxy]-1-oxa-6-azacyclopentadecan-15-o |
InChI Key | MQTOSJVFKKJCRP-OHJWJPDZSA-N |
SMILES | CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)C(C)[C@@H](O[C@@H]2O[C@H](C)C[C@@H]([C@H]2O)N(C)C)[C@](C)(O)C[C@@H](C)CN(C)[C@H](C)[C@@H](O)[C@]1(C)O |
IUPAC Name | (2R,3S,4R,5R,8R,10R,11R,13S,14R)-11-{[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy}-2-ethyl-3,4,10-trihydroxy-13-{[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy}-3,5,6,8,10,12,14-heptamethyl-1-oxa-6-azacyclopentadecan-15-one |
Molecular Weight (g/mol) | 749.00 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'