Learn More
Azlon™ Acrylic Single-Bore Class B Buret
Suitable for left or right handed use. The Azlon™ Acrylic Single Bore Class B Buret is great for educational laboratories, especially where breakages often occur.
Brand:  Azlon BDH112T
 View more versions of this product
Product Code. 12044637
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Suitable for left or right handed use. The Azlon™ Acrylic Single Bore Class B Buret is great for educational laboratories, especially where breakages often occur.
  • Acrylic construction for strength and transparency
  • Graduations are screen printed in black to ISO 385 Class B accuracy
  • Suitable for use in educational laboratories especially where breakages often occur
  • Unaffected by most dilute aqueous solutions, however unsuitable for use with organic solvents
  • Turning the stopcock through 90° makes it suitable for left or right handed use
  • Stopcocks can be removed for easy cleaning
TRUSTED_SUSTAINABILITY
Specifications
Specifications
25 mL
0.1 mL
PMP/Polyethylene Stopcocks
±0.1 mL
Class B
Acrylic
Educational Laboratories Especially where Breakages Often Occur
PMP, PTFE
Single Bore Buret