Learn More
Azlon™ Ampoule Breaker
Break glass ampoules safely with easy and safe disposable of the broken glass . The Azlon™ ampoule breaker comes in a choice of sizes.
Brand: Azlon AWY702
630.14 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Enables safe and easy breaking of glass ampoules
- Broken ampoule top is retained for safe disposal
- Choice of size to suit all common glass ampoules
Specifications
Natural | |
Ampule Breaker | |
HDPE | |
5-10 mL Ampoule |