missing translation for 'onlineSavingsMsg'
Learn More

Azlon™ Square Shaped HDPE Bucket

Square shaped buckets suitable for food contact applications in accordance with EU regulations

Brand:  Azlon BWX116

37.72 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 12075562

  • 40.70 € / Each
Estimated Shipment: 01-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Durable square shape allows economical use of space
  • Handle grip for easy lifting and carrying
  • Moulded graduations (14 litre x 0.5 divisions)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Square Bucket
Not Autoclavable
Polyethylene
292 mm
292 mm
HDPE
White
292 mm
1 Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.