missing translation for 'onlineSavingsMsg'
Learn More

Azlon™ Industrial HDPE Funnel

Constructed of Polyethylene and unbreakable in normal use. Azlon™ Industrial HDPE Funnels are suitable for food contact applications in accordance with EU and USA regulations. Feature straight sided upper rims to prevent spillage.

Brand:  Azlon FWC106

 View more versions of this product

Product Code. 11373664

  • 50.05 € / Pack of 10
Estimated Shipment: 30-05-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

Constructed of Polyethylene and unbreakable in normal use. Azlon™ Industrial HDPE Funnels are suitable for food contact applications in accordance with EU and USA regulations. Feature straight sided upper rims to prevent spillage.

  • Rigid industrial funnels in polyethylene
  • Unbreakable in normal use
  • Feature straight sided upper rims to prevent spillage
TRUSTED_SUSTAINABILITY
Specifications

Specifications

120mm
12mm
121 mm
450 mL
HDPE
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Azlon™ Industrial HDPE Funnel > Height: 121mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.