missing translation for 'onlineSavingsMsg'
Learn More

Azlon™ LDPE DIY Printed Safety Wash Bottles

Product Code. 12530066
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
Closure Type:
Non-venting Cap
Vapor-venting
Closure Color:
Assorted
White
This item is not returnable. View return policy
This item is not returnable. View return policy

Create custom labels for these do-it-yourself LDPE wash bottles. Azlon™ LDPE DIY Printed Safety Wash Bottles come with adhesive hazard labels and are ideal for a variety of liquid storage and sampling applications. DripLok™ vapor venting caps provide increased safety with volatile solvents.

  • Do-it-yourself wash bottles with blank write-on panel
  • Indelibly printed in three languages (English, French and German)
  • Supplied with adhesive hazard labels
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 1000 mL
Color Translucent
Diameter (Metric) Outer Neck 38 mm
Diameter (Metric) Inner Neck 34 mm
Includes Closure, blank write-on panel, adhesive hazard signs
Material LDPE
Quantity 5/PK
Temperature Range (Metric) –50 to 80°C
Closure Color White
Closure Included Yes
Autoclavable Not Autoclavable
Mouth Wide
Height (Metric) 202 mm
Closure Material Polypropylene
Diameter (Metric) 90 mm
Closure Type Non-venting Cap
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.