Learn More
Azlon™ LDPE Printed Slope Shoulder Wash Bottles
Use these durable plastic bottles for a wide range of liquid storage, media preparation and sampling applications. Azlon™ LDPE Printed Slope Shoulder Wash Bottles are indelibly printed with a choice of seven popular laboratory solvents, appropriate hazard symbols and CAS numbers.
Brand: Azlon™ WGF601P
34.58 EUR valid until 2025-03-30
Use promo code "24134" to get your promotional price.
Description
Use these durable plastic bottles for a wide range of liquid storage, media preparation and sampling applications. AzlonTM LDPE Printed Slope Shoulder Wash Bottles are indelibly printed with a choice of seven popular laboratory solvents, appropriate hazard symbols and CAS numbers.
- Labeled wash bottles for easy identification of contents
- Slim design enables comfortable handling and enhanced ease of use
- Fitted with color-coded polypropylene closures
- Closures feature precision jet-tip aperture for fine control
Specifications
500 mL | |
Translucent | |
For easy identification of contents | |
LDPE | |
White | |
Not Autoclavable | |
Narrow | |
187 mm | |
Distilled Water | |
187 mm | |
Yes |
16.91 oz. | |
69mm | |
White Polypropylene closure | |
5/PK | |
Yes | |
No | |
Round | |
Wash Bottle | |
Polypropylene | |
Cap |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.