missing translation for 'onlineSavingsMsg'
Learn More

Azlon™ Pivot Ring Magnetic Stirrer Bar

Constructed of magnetic PTFE. Azlon™ Pivot Ring Stirrer Bar includes a central pivot ring that ensures efficient spinning even on curved or uneven bases.

Brand:  Azlon SWN504

9.91 EUR valid until 2024-11-30
Use promo code "21840" to get your promotional price.


 View more versions of this product

Product Code. 10135374

  • 11.65 € / Pack of 2
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

Constructed of magnetic PTFE. Azlon™ Pivot Ring Stirrer Bar includes a central pivot ring that ensures efficient spinning even on curved or uneven bases.

  • Constructed of magnetic PTFE
  • Central pivot ring ensures efficient spinning even on curved or uneven bases
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Autoclavable
0.98 in.
6 mm
Magnet
Pivot ring
0.98 Lin.
PTFE
25 mm
2/Pk.
Stir Bar
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers