Learn More
Azlon™ PMP Autoclavable Tweezer
Manufactured with PMP which has a high melting point and good temperature stability. Azlon™ PMP Autoclavable Tweezers are ideal for secure handling of samples.
4.90 € - 7.75 €
Specifications
Tip Style | Sharp |
---|---|
Material | PMP |
Product Type | Tweezers |
For Use With (Application) | For secure handling of samples |
Description
Manufactured with PMP which has a high melting point and good temperature stability. Azlon™ PMP Autoclavable Tweezers are ideal for secure handling of samples.
- High quality, autoclavable tweezers
- Sharp tip
- Fine tooth grips for secure handling of samples
Specifications
Sharp | |
PMP | |
Tweezers | |
For secure handling of samples |