missing translation for 'onlineSavingsMsg'
Learn More

Azlon™ Polycarbonate Hinged Safety Shield

Protect yourself from potential hazards with a light weight but very strong safety shield. The Azlon™ polycarbonate hinged safety shield can be secured to a lab bench.

560.00 € - 1180.00 €

Specifications

Material Polycarbonate
Color Clear
For Use With (Application) For excellent protection in potentially hazardous situations
No. per Pack 1EA
Quantity 1EA
View More Specs

Products 3
Product Code Brand Width (Metric) Length (Metric) Price Quantity & Availability  
Product Code Brand Width (Metric) Length (Metric) Price Quantity & Availability  
10748955
View Documents
Azlon
SGT002
590 mm 38 cm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10093294
View Documents
Azlon
SGT004
590 mm 61 cm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10606855
View Documents
Azlon
SGT006
750 mm 61 cm
1180.00 €
Each
Estimated Shipment: 03-02-2025
to see stock.
Add to basket
 
Description

Description

  • Excellent protection in potentially hazardous situations
  • Adjustable - can be used as a tight triangle or extended out to a straight line configuration
  • Incredibly strong and lightweight
  • Supplied with two pairs of plastic coated metal feet
  • Feet may be clamped or bolted onto the bench to give greater stability
Specifications

Specifications

Polycarbonate
For excellent protection in potentially hazardous situations
1EA
Clear
1EA
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.