missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azlon™ Polypropylene Measuring Scoop
Click to view available options
Length (Metric):
60 mm
82 mm
102 mm
137 mm
160 mm
198 mm
259 mm
315 mm
385 mm
Capacity (Metric):
2 mL
5 mL
10 mL
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
missing translation for 'unitSize'
Each
Pack of 5
Beschreibung
Scoop chemical reagents or food items safely. The Azlon™ polypropylene measuring scoop comes with flat top and bottom.
- Made from food safe polypropylene
- Top is flat for levelling off powders
- Base is flat enabling scoops to stand horizontally
- The capacity is clearly molded on each size
Spezifikation
Spezifikation
| Product Type | Measuring Scoop |
| Material | Polypropylene |
| Includes | Handle |
| Capacity (Metric) | 500 mL |
| Sterility | Non-sterile |
| Length (Metric) | 315 mm |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur