missing translation for 'onlineSavingsMsg'
Learn More

Azlon™ PTFE Sleeves

Fabricated from pure PTFE for superior chemical, corrosion and contamination resistance, high thermal stability and good flexural properties. Azlon™ PTFE Sleeves form a grease-free gas tight seal for ground glass joints. Available in standard joint sizes for accurate fit.

Brand:  Azlon SWN967

 View more versions of this product

Product Code. 12541456

  • 58.95 € / Pack of 10
Estimated Shipment: 17-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Fabricated from pure PTFE for superior chemical, corrosion and contamination resistance, high thermal stability and good flexural properties. Azlon™ PTFE Sleeves form a grease-free gas tight seal for ground glass joints. Available in standard joint sizes for accurate fit.

  • Truncated cone of pure PTFE
  • Allows easy separation of joints
  • Excellent chemical resistance
  • Outstanding resistance to fatigue
  • Working temperature range of -100°C to +250°C
  • No contamination from joint grease
  • Intended for single or occasional use
  • Appropriate for low vacuum work
  • USP Class VI grade PTFE
  • Verified for food contact applications per US regulations
TRUSTED_SUSTAINABILITY
Specifications

Specifications

For low vacuum work
Sleeves
45/50
PTFE
–100 to 250°C
Truncated cone
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.