missing translation for 'onlineSavingsMsg'
Learn More

Azlon™ Rubber Pipette Fillers

Suitable for pipettes up to 100mL

Brand:  Azlon PWY146

25.12 EUR valid until 2025-03-30
Use promo code "24134" to get your promotional price.


 View more versions of this product

Product Code. 15718047

  • 28.85 € / Each
Estimated Shipment: 17-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Suitable for pipettes up to 100mL

  • Suitable for pipettes up to 100mL
  • Pipette filler works via a series of glass ball valves for air release, suction and delivery
  • PWY145 is suitable for single handed operation
  • PWY146 has an extended insert nozzle for improved grip
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Rubber
Pipette Filler
1/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.