Learn More
Azlon™ Spare Stopcock for HDPE Aspirator
Replaces leaking or worn out stopcocks. Azlon™ Spare Stopcock for HDPE Aspirators create an excellent seal and are leakproof when installed properly.
Brand:  Azlon™ SCBNP01
24.13 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
Product Code. 11333464
  • 26.05 € / Each
Estimated Shipment: 23-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For use with Heavy Duty Polypropylene (HDPE) Aspirators
  • Opens and closes in a quarter turn
  • Leakproof
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Spare Stopcock
HDPE
HDPE aspirator bottle