Learn More
Synergy Healthcare™ Azo™ Flat Pouches
High quality preformed, heat sealable, peelable sterile barrier packaging. Synergy Healthcare™ Azo™ Flat Pouches are suited to the sterilization of surgical instruments, laboratory equipment or small to medium sized sets using steam or Ethylene Oxide.
Brand:  Synergy Healthcare™ MBO1013
 View more versions of this product
Product Code. 12769005
  • 779.00 € / Pack of 1000
Estimated Shipment: 28-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Made from specially treated paper, less fibres released when pouch is opened and reducing the risk of contamination
  • New film technology increases impact strength of pouch film up to 10 times, virtually eliminates the shattering of film when opening
  • Indicators are placed between the 2 layers which make-up the pouch film, prevents offsetting and leakage of ink
  • Meet EN ISO 11607 part 1 and EN 868-4 standards
Specifications
Specifications
Cool dry place
EN868-5, ISO11607-1, 93/42/EEC, ISO13485, ISO9001
Blue
Polyethylene
135 μm
170°C to 210°C
420 x 200mm
200 mm
Autoclavable
Heat Seal
Single-use
Non-sterile
1200
6.30 kN/m
420 mm