Learn More
Synergy Healthcare™ Azo™ Flat Reel Sterilizer Bags
High quality preformed, heat sealable, peelable sterile barrier packaging. Synergy Healthcare™ Azo™ Flat Reel Sterilizer Bags are suited to the sterilization of surgical instruments, laboratory equipment or small to medium sized sets using steam or Ethylene Oxide.
Brand:  Synergy Healthcare™ RF100200
 View more versions of this product
Product Code. 15601974
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Protects its contents from deterioration, damage and contamination
  • Meet EN ISO 11607 part 1 and EN 868-4 standards
Specifications
Specifications
Autoclave Bag
EN868-5 and ISO11607-1
Blue
Polyethylene
4 reels
200 m
Autoclavable
Heat Seal
Single-use
Non-sterile
200m x 100mm
100 mm