missing translation for 'onlineSavingsMsg'
Learn More
Azobenzene, 97+%, Thermo Scientific Chemicals Product Code.: 11488623

Azobenzene, 97+%, Thermo Scientific Chemicals

Product Code. 11488623
25 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11488623

Brand: Thermo Scientific Alfa Aesar A10425.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Azobenzene is used to construct photoswitch tethered ligands to turn protein activity on and off. It is also used to increase the extracellular or intracellular concentration of active molecules to study the mechanism of action of sympathetic and parasympathetic neuromediators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Azobenzene is used to construct photoswitch tethered ligands to turn protein activity on and off. It is also used to increase the extracellular or intracellular concentration of active molecules to study the mechanism of action of sympathetic and parasympathetic neuromediators.

Solubility
Soluble in alcohol, ether, benzene and glacial acetic acid. Insoluble in water.

Notes
Store at 4°C. Incompatible with strong oxidizing agents. Air and light sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-33-3
Molecular Formula C12H10N2
Molecular Weight (g/mol) 182.23
MDL Number MFCD00003022
InChI Key DMLAVOWQYNRWNQ-UHFFFAOYSA-N
Synonym azobenzene, diazene, diphenyl, diazobenzene, e-diphenyldiazene, azobisbenzene, azofume, 1,2-diphenyldiazene, azodibenzeneazofume, diphenyldiimide, azobenzideShow MoreShow Less
PubChem CID 2272
ChEBI CHEBI:190358
IUPAC Name diphenyldiazene
SMILES C1=CC=C(C=C1)N=NC1=CC=CC=C1

Specifications

Melting Point 65°C to 69°C
Density 1.203
Boiling Point 293°C
Quantity 25 g
UN Number UN3077
Merck Index 14,917
Solubility Information Soluble in alcohol,ether,benzene and glacial acetic acid. Insoluble in water.
Formula Weight 182.23
Percent Purity ≥97%
Chemical Name or Material Azobenzene

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10210582 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10425.14,A10425.36,A10425.22
Lot NumberC4182C Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10425.14,A10425.36,A10425.22
Lot Number10211752 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10425.14,A10425.36,A10425.22
Lot NumberA5944B Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10425.14,A10425.36,A10425.22
Lot Number10183909 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10425.14,A10425.36,A10425.22
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P281-Use personal protective equipment as required.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Azobenzene, 97+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.