missing translation for 'onlineSavingsMsg'
Learn More

Synergy Healthcare™ Azowipe™ Hard Surface Disinfecting Wipes

Product Code. p-8647001
Click to view available options
Dimensions (L x W):
20 x 18 cm
Unit Size:
Pack of 200
This item is not returnable. View return policy

Product Code. 15649795

Brand: Synergy Healthcare™ 81103

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Provides rapid disinfection of hard surfaces. Synergy Healthcare™ Azowipe™ Hard Surface Disinfectant Wipes are non-smearing, strong and durable for everyday use.

  • 70% Isopropyl alcohol disinfectant
  • Broad spectrum activity against a range of micro-organisms
  • Extremely fast contact times from 15 seconds
  • Non-smearing and streak-free when used on hard surfaces.
  • Suitable for food manufacturing, hospitality and catering sectors.
  • Compliant with HTM-01-05 guidelines

Specifications

Product Type Disinfectant Wipe
Length (Metric) 20 cm
Width (Metric) 18 cm
Certifications/Compliance EN1276, EN13727, EN14476, EN1650/EN13624, EN14348
Disposable Single-use
Dimensions (L x W) 20 x 18 cm
Format Wiper
Packaging Quantity 200 Wipes/Dispenser
Saturation Solution 70% isopropyl alcohol
compliance-icons compliance-icons
Product Identifier
  • azowipe
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
Supplemental information
  • MIXTURE LIST-Contains : propan-2-ol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.