missing translation for 'onlineSavingsMsg'
Learn More

Azpack™ Mortar and Pestle with Wooden Handle Pestle

Releases oils that can't be extracted using modern appliances. Azpack™ Mortar and Pestle with Wooden Handle will grind and crush the toughest materials without cracking the strong and dense porcelain.

Brand:  Azpack™ 12303348

 View more versions of this product

Product Code. 12303348

  • 131.00 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

You may only order up to the available stock. Where an alternative item has been identified it will be shown below
This item is not returnable. View return policy

Description

Description

  • Mortar constructed of vitrified electical porcelain
  • Pestle includes a Beech wood handle
  • Unglazed grinding surface
  • Integral pouring lip reduces waste and spillage
Specifications

Specifications

284 mL
107mm
Mortar and Pestle
72mm (Mortar)
Unglazed
129mm (135mm to lip)
Mortar: Vitrified electrical porcelain, Pestle: Beech
192mm (Pestle)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.