missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ B-27™ Supplement (50X), custom
Click to view available options
Quantity:
10 mL
Unit Size:
Each
Description
The custom B-27™ Supplement (50X) is a serum-free additive for the growth and expansion of retinal ganglion cells. It is supplied as a 50X liquid concentrate and it is used with a basal medium, such as Neurobasal™, and other additives, such as GlutaMAX-1.
Specifications
Specifications
| Content And Storage | 10 ml - 50X frozen concentrate; store at -5 to -20°C in the dark. |
| Cell Type | Neural Cells |
| Classification | Serum-free |
| Concentration | 50 X |
| Culture Type | Mammalian Cell Culture |
| Form | Liquid |
| Product Type | Cell Culture Supplement |
| Serum Level | Serum-Free |
| Sterility | Sterile |
| With Additives | Antioxidant Cocktail, Insulin, Vitamin A |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction